Skip to Content
MilliporeSigma
DPPF Pd G3 95%

DPPF Pd G3

DPPF Pd G3 95%
Synonym(s):
Methanesulfonato 1,1-ferrocenediyl-bis(diphenylphosphino) (2′-amino-1,1′-biphenyl-2-yl) palladium(II)
Empirical Formula (Hill Notation):
C47H41FeNO3P2PdS
CAS Number:
Molecular Weight:
924.11

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing