Skip to Content
MilliporeSigma
DMT-2′O-Methyl-rU Phosphoramidite configured for ABI

DMT-2′O-Methyl-rU Phosphoramidite

DMT-2′O-Methyl-rU Phosphoramidite configured for ABI
Synonym(s):
5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-methyl-uridine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite], DMT-2′O-Methyl-rU amidite
Empirical Formula (Hill Notation):
C40H49N4O9P
CAS Number:
Molecular Weight:
760.81

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Technique