Skip to Content
MilliporeSigma
(+)-Magnesium L-ascorbate ≥90% (T)

(+)-Magnesium L-ascorbate

(+)-Magnesium L-ascorbate ≥90% (T)
Synonym(s):
L(+)-Ascorbic acid magnesium salt, Vitamin C magnesium salt
Empirical Formula (Hill Notation):
C12H14MgO12
CAS Number:
Molecular Weight:
374.54
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing