Skip to Content
MilliporeSigma
DL-Tartaric acid ReagentPlus®, 99%

DL-Tartaric acid

DL-Tartaric acid ReagentPlus®, 99%
Synonym(s):
DL-2,3-Dihydroxybutanedioic acid, DL-Tartaric acid
Linear Formula:
HOOC(CHOH)2COOH
CAS Number:
Molecular Weight:
150.09

Products

Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Organoleptic Class
Showing 1-3 of 3
Compare
Product Number
Description
Pricing