Skip to Content
MilliporeSigma
DL-Isoserine 98%

DL-Isoserine

DL-Isoserine 98%
Synonym(s):
(±)-3-Amino-2-hydroxypropionic acid
Linear Formula:
H2NCH2CH(OH)CO2H
CAS Number:
Molecular Weight:
105.09

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing