Skip to Content
MilliporeSigma
(±)-Cotinine-d3 solution 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

(±)-Cotinine-d3 solution

(±)-Cotinine-d3 solution 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Synonym(s):
(+/-)-Cotinine-d3 solution
Empirical Formula (Hill Notation):
C10H9D3N2O
CAS Number:
Molecular Weight:
179.23
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Format
Application
Package Size
Reference Material Type
Special Grade
Technique