Skip to Content
MilliporeSigma
D-Lactitol monohydrate ≥99% (HPLC)

D-Lactitol monohydrate

D-Lactitol monohydrate ≥99% (HPLC)
Synonym(s):
4-O-β-D-Galactopyranosyl-D-glucitol
Empirical Formula (Hill Notation):
C12H24O11 · H2O
CAS Number:
Molecular Weight:
362.33
EC Number:

Products

Product Category
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing