Skip to Content
MilliporeSigma
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid 97%

(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid

(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid 97%
Synonym(s):
Trolox
Empirical Formula (Hill Notation):
C14H18O4
CAS Number:
Molecular Weight:
250.29
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing