Skip to Content
MilliporeSigma
Di-μ-chloro-tetracarbonyldirhodium(I) 97%

Di-μ-chloro-tetracarbonyldirhodium(I)

Di-μ-chloro-tetracarbonyldirhodium(I) 97%
Synonym(s):
Rhodium carbonyl chloride(I), Rhodium(I) carbonyl chloride, Rhodium(I) dicarbonyl chloride dimer, Tetracarbonyldi-μ−chlorodirhodium(I)
Linear Formula:
[Rh(CO)2Cl]2
CAS Number:
Molecular Weight:
388.76
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing