Skip to Content
MilliporeSigma
Di-tert-butyl N,N-diisopropylphosphoramidite 95%

Di-tert-butyl N,N-diisopropylphosphoramidite

Di-tert-butyl N,N-diisopropylphosphoramidite 95%
Linear Formula:
[(CH3)2CH]2NP[OC(CH3)3]2
CAS Number:
Molecular Weight:
277.38

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing