![di-Potassium hydrogen phosphate trihydrate buffer substance for chromatography LiChropur™](/deepweb/assets/sigmaaldrich/product/images/416/076/53566e58-8fe6-49fb-904e-a2cea87aa289/640/53566e58-8fe6-49fb-904e-a2cea87aa289.jpg)
di-Potassium hydrogen phosphate trihydrate
![di-Potassium hydrogen phosphate trihydrate buffer substance for chromatography LiChropur™](/deepweb/assets/sigmaaldrich/product/images/416/076/53566e58-8fe6-49fb-904e-a2cea87aa289/640/53566e58-8fe6-49fb-904e-a2cea87aa289.jpg)
Synonym(s):
di-Potassium hydrogen phosphate trihydrate
Empirical Formula (Hill Notation):
HK2O4P · 3H2O
CAS Number:
Molecular Weight:
228.22
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Technique
Showing 1-2 of 2