![di-Potassium hydrogen phosphate anhydrous low in sodium, EMPROVE® API, Ph. Eur., BP, USP](/deepweb/assets/sigmaaldrich/product/structures/326/934/a5186a5a-998c-403f-884c-70701cd60ca9/640/a5186a5a-998c-403f-884c-70701cd60ca9.png)
di-Potassium hydrogen phosphate anhydrous
![di-Potassium hydrogen phosphate anhydrous low in sodium, EMPROVE® API, Ph. Eur., BP, USP](/deepweb/assets/sigmaaldrich/product/structures/326/934/a5186a5a-998c-403f-884c-70701cd60ca9/640/a5186a5a-998c-403f-884c-70701cd60ca9.png)
Synonym(s):
Potassium phosphate dibasic, Dipotassium hydrogenphosphate, Dipotassium phosphate, sec.-Potassium phosphate
Linear Formula:
K2HPO4
CAS Number:
Molecular Weight:
174.18
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing