Skip to Content
CA
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
mRNA Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
DIPHENYL(VINYL)PHOSPHINE OXIDE
Linear Formula:
C
14
H
13
OP
CAS Number:
2096-78-8
Molecular Weight:
228.233
Products
Formula Weight
Special Grade
Aldrich
CPR
(3)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
S80749
DIPHENYL(VINYL)PHOSPHINE OXIDE,
Aldrich
CPR
Expand
View Pricing
S326658
DIPHENYL(VINYL)PHOSPHINE OXIDE,
Aldrich
CPR
Expand
View Pricing
S322954
DIPHENYL(VINYL)PHOSPHINE OXIDE,
Aldrich
CPR
Expand
View Pricing