Skip to Content
MilliporeSigma
Dimidium bromide BioReagent, suitable for fluorescence, ~95% (AT)

Dimidium bromide

Dimidium bromide BioReagent, suitable for fluorescence, ~95% (AT)
Synonym(s):
3,8-Diamino-5-methyl-6-phenylphenanthridinium bromide, Trypadine, Dimidium bromide
Empirical Formula (Hill Notation):
C20H18BrN3
CAS Number:
Molecular Weight:
380.28

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Chemical Composition
Quality Segment
Showing 1-2 of 2