Skip to Content
MilliporeSigma
Dimethyl trisulfide analytical standard

Dimethyl trisulfide

Dimethyl trisulfide analytical standard
Linear Formula:
CH3SSSCH3
CAS Number:
Molecular Weight:
126.26
EC Number:

Products

Product Category
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing