Skip to Content
MilliporeSigma
Dimethyl suberimidate dihydrochloride 98%

Dimethyl suberimidate dihydrochloride

Dimethyl suberimidate dihydrochloride 98%
Synonym(s):
DMS, Dimethyl ester octanediimidic acid dihydrochloride
Linear Formula:
CH3OC(=NH)CH2(CH2)4CH2C(=NH)OCH3 · 2HCl
CAS Number:
Molecular Weight:
273.20
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing