Skip to Content
MilliporeSigma
Dimethylditetradecylammonium bromide ≥97.0% (NT)

Dimethylditetradecylammonium bromide

Dimethylditetradecylammonium bromide ≥97.0% (NT)
Synonym(s):
Ditetradecyldimethylammonium bromide
Linear Formula:
[CH3(CH2)13]2N(Br)(CH3)2
CAS Number:
Molecular Weight:
518.74
EC Number:

Products

Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing