Skip to Content
MilliporeSigma
Diisopropylsilyl bis(trifluoromethanesulfonate) 96%

Diisopropylsilyl bis(trifluoromethanesulfonate)

Diisopropylsilyl bis(trifluoromethanesulfonate) 96%
Synonym(s):
Trifluoromethanesulfonic acid diisopropylsilylene ester
Linear Formula:
(CF3SO3)2Si[CH(CH3)2]2
CAS Number:
Molecular Weight:
412.40

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing