Skip to Content
MilliporeSigma
Dihydroxyacetone phosphate dilithium salt ≥93% dry basis (enzymatic)

Dihydroxyacetone phosphate dilithium salt

Dihydroxyacetone phosphate dilithium salt ≥93% dry basis (enzymatic)
Synonym(s):
1,3-Dihydroxy-2-propanone 1-phosphate dilithium salt, 1-Hydroxy-3-(phosphonooxy)-2-propanone dilithium salt, DHAP
Empirical Formula (Hill Notation):
C3H5Li2O6P
CAS Number:
Molecular Weight:
181.92

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing