Skip to Content
MilliporeSigma
Dihydromyrcenol ≥99%

Dihydromyrcenol

Dihydromyrcenol ≥99%
Linear Formula:
H2C=CHCH(CH3)(CH2)3C(CH3)2OH
CAS Number:
Molecular Weight:
156.27
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Application
Markush Class
Markush Group
Organoleptic Class
Showing 1-1 of 1
Compare
Product Number
Description
Pricing