Skip to Content
MilliporeSigma
Dihydrocapsaicin pharmaceutical secondary standard, certified reference material

Dihydrocapsaicin

Dihydrocapsaicin pharmaceutical secondary standard, certified reference material
Synonym(s):
6,7-Dihydrocapsaicin, 8-Methyl-N-vanillylnonanamide, N-[(4-Hydroxy-3-methoxyphenyl)methyl]-8-methylnonenamide, Dihydrocapsaicin
Empirical Formula (Hill Notation):
C18H29NO3
CAS Number:
Molecular Weight:
307.43

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique