Skip to Content
MilliporeSigma
Digoxigenin NHS-ester ≥80% (HPLC)

Digoxigenin NHS-ester

Digoxigenin NHS-ester ≥80% (HPLC)
Synonym(s):
ε-(Digoxigenin-3-0-acetamido)caproic acid N-hydroxysuccinimide ester, DIG NHS
Empirical Formula (Hill Notation):
C35H50N2O10
CAS Number:
Molecular Weight:
658.78

Products

Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing