![Diflunisal analytical standard](/deepweb/assets/sigmaaldrich/product/structures/427/633/53a6b91b-3032-4b2b-bd40-ce2ea1794ae8/640/53a6b91b-3032-4b2b-bd40-ce2ea1794ae8.png)
Diflunisal
![Diflunisal analytical standard](/deepweb/assets/sigmaaldrich/product/structures/427/633/53a6b91b-3032-4b2b-bd40-ce2ea1794ae8/640/53a6b91b-3032-4b2b-bd40-ce2ea1794ae8.png)
Synonym(s):
Diflunisal, 2′,4′-Difluoro-4-hydroxy-[1,1′-biphenyl]-3-carboxylic acid, 5-(2,4-Difluorophenyl)salicylic acid
Empirical Formula (Hill Notation):
C13H8F2O3
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing