Skip to Content
MilliporeSigma
Diethylphosphonoacetic acid 95%

Diethylphosphonoacetic acid

Diethylphosphonoacetic acid 95%
Synonym(s):
(Diethoxyphosphinyl)acetic acid, Diethyl carboxymethylphosphonate, NSC 272281
Linear Formula:
(C2H5O)2P(O)CH2CO2H
CAS Number:
Molecular Weight:
196.14

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing