Skip to Content
MilliporeSigma
Diethylenetriamine-pentaacetic acid calcium trisodium salt hydrate ≥97.0% (dried material, KT)

Diethylenetriamine-pentaacetic acid calcium trisodium salt hydrate

Diethylenetriamine-pentaacetic acid calcium trisodium salt hydrate ≥97.0% (dried material, KT)
Synonym(s):
Calcium-trisodium diethylenetriamine-pentaacetate hydrate
Empirical Formula (Hill Notation):
C14H18CaN3Na3O10 · xH2O
CAS Number:
Molecular Weight:
497.35 (anhydrous basis)

Products

Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1