Skip to Content
MilliporeSigma
Dicycloverine for system suitability European Pharmacopoeia (EP) Reference Standard

Dicycloverine for system suitability

Dicycloverine for system suitability European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Dicyclomine hydrochloride, 2-(Diethylamino)ethyl 1-cyclohexylcyclohexane-1-carboxylate hydrochloride, [1,1-Bicyclohexyl]-1-carboxylic acid 2-(diethylamino)ethyl ester hydrochloride
Empirical Formula (Hill Notation):
C19H35NO2 · HCl
CAS Number:
Molecular Weight:
345.95
EC Number:

Products

Formula Weight
API Family
Reference Material Type
Showing 1-1 of 1