Skip to Content
MilliporeSigma
Diclofenac sodium salt analytical standard

Diclofenac sodium salt

Diclofenac sodium salt analytical standard
Synonym(s):
2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid sodium salt
Empirical Formula (Hill Notation):
C14H10Cl2NNaO2
CAS Number:
Molecular Weight:
318.13
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique