Skip to Content
MilliporeSigma
Dichlorodimethylsilane solution ~5% in toluene, LiChropur™

Dichlorodimethylsilane solution

Dichlorodimethylsilane solution ~5% in toluene, LiChropur™
Synonym(s):
Dimethyldichlorosilane solution
Empirical Formula (Hill Notation):
C2H6Cl2Si
CAS Number:
Molecular Weight:
129.06

Products

Product Category
Formula Weight
Physical Form
Quality Segment
Reaction Type
Reagent Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing