Skip to Content
MilliporeSigma
Dichloro(1,5-cyclooctadiene)ruthenium(II), polymer 95%

Dichloro(1,5-cyclooctadiene)ruthenium(II), polymer

Dichloro(1,5-cyclooctadiene)ruthenium(II), polymer 95%
Synonym(s):
(1,5-Cyclooctadiene)ruthenium(II) chloride, Ruthenium(II) chloride 1,5-cyclooctadiene complex
Linear Formula:
[Ru(COD)Cl2]n
CAS Number:
Molecular Weight:
280.16 (as monomer)

Products

Brand
Sustainability
Feature
Formula Weight
Purity
Chain Type
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing