Dibutylammonium acetate solution
Synonym(s):
Dibutylamine acetate Concentrate
Linear Formula:
(CH3CH2CH2CH2)2NH · CH3COOH
CAS Number:
Molecular Weight:
189.30
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Package Size
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing