Skip to Content
MilliporeSigma
Dibenzyl phosphite technical grade

Dibenzyl phosphite

Dibenzyl phosphite technical grade
Synonym(s):
Dibenzyl phosphonate
Linear Formula:
(C6H5CH2O)2P(O)H
CAS Number:
Molecular Weight:
262.24
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing