Skip to Content
MilliporeSigma
Dibenzo-18-crown-6 98%

Dibenzo-18-crown-6

Dibenzo-18-crown-6 98%
Synonym(s):
2,3,11,12-Dibenzo-1,4,7,10,13,16-hexaoxacyclooctadeca-2,11-diene, 6,7,9,10,17,18,20,21-Octahydrodibenzo[b,k][1,4,7,10,13,16]hexaoxacyclooctadecene, DB18C6
Empirical Formula (Hill Notation):
C20H24O6
CAS Number:
Molecular Weight:
360.40
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing