Skip to Content
MilliporeSigma
di-Ammonium oxalate monohydrate EMPLURA®

di-Ammonium oxalate monohydrate

di-Ammonium oxalate monohydrate EMPLURA®
Synonym(s):
di-Ammonium oxalate monohydrate
Linear Formula:
(NH4)2C2O4 · H2O
CAS Number:
Molecular Weight:
124.10 (anhydrous basis)

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Showing 1-2 of 2