Skip to Content
MilliporeSigma
Di(2-pyridyl) thionocarbonate 98%

Di(2-pyridyl) thionocarbonate

Di(2-pyridyl) thionocarbonate 98%
Empirical Formula (Hill Notation):
C11H8N2O2S
CAS Number:
Molecular Weight:
232.26

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing