Skip to Content
MilliporeSigma
(DHQ)2Pyr 97%

(DHQ)2Pyr

(DHQ)2Pyr 97%
Synonym(s):
Hydroquinine 2,5-diphenyl-4,6-pyrimidinediyl diether
Empirical Formula (Hill Notation):
C56H60N6O4
CAS Number:
Molecular Weight:
881.11

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing