Skip to Content
MilliporeSigma
(DHQ)2PHAL ≥95%

(DHQ)2PHAL

(DHQ)2PHAL ≥95%
Synonym(s):
Hydroquinine 1,4-phthalazinediyl diether
Empirical Formula (Hill Notation):
C48H54N6O4
CAS Number:
Molecular Weight:
778.98

Products

Brand
Formula Weight
Melting Point (°C)
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing