Skip to Content
MilliporeSigma
D-Galacturonic acid sodium salt ≥95.0% (T)

D-Galacturonic acid sodium salt

D-Galacturonic acid sodium salt ≥95.0% (T)
Empirical Formula (Hill Notation):
C6H9NaO7
CAS Number:
Molecular Weight:
216.12

Products

Product Category
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Optical Isomer
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing