Skip to Content
MilliporeSigma
Dextromethorphan hydrobromide monohydrate ≥99% (TLC)

Dextromethorphan hydrobromide monohydrate

Dextromethorphan hydrobromide monohydrate ≥99% (TLC)
Synonym(s):
(9S,13S,14S)-3-Methoxy-17-methylmorphinan hydrobromide
Empirical Formula (Hill Notation):
C18H25NO · HBr · H2O
CAS Number:
Molecular Weight:
370.32

Products

Product Category
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing