
Dextromethorphan for system suitability CRS

Synonym(s):
Dextromethorphan hydrobromide monohydrate, (9S,13S,14S)-3-Methoxy-17-methylmorphinan hydrobromide
Empirical Formula (Hill Notation):
C18H25NO · HBr · H2O
CAS Number:
Molecular Weight:
370.32
Products
Formula Weight
Manufacturer
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing