Skip to Content
MilliporeSigma
Dexamethasone British Pharmacopoeia (BP) Assay Standard

Dexamethasone

Dexamethasone British Pharmacopoeia (BP) Assay Standard
Synonym(s):
(11β,16α)-9-Fluoro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione, 9α-Fluoro-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, 9α-Fluoro-16α-methylprednisolone, Prednisolone F, Dexamethasone
Empirical Formula (Hill Notation):
C22H29FO5
CAS Number:
Molecular Weight:
392.46
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique