Skip to Content
MilliporeSigma
vibrant-m

Dermatan sulfate

vibrant-m
Synonym(s):
Chondroitin sulfate B sodium salt, β-Heparin, Dermatan sulfate sodium salt
CAS Number:

Products

Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing