Skip to Content
MilliporeSigma
D-Cysteine hydrochloride monohydrate ≥98% (TLC)

D-Cysteine hydrochloride monohydrate

D-Cysteine hydrochloride monohydrate ≥98% (TLC)
Linear Formula:
HSCH2CH(NH2)COOH · HCl · H2O
CAS Number:
Molecular Weight:
175.63
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Application
Optical Isomer
Amino Acid
Showing 1-1 of 1
Compare
Product Number
Description
Pricing