Skip to Content
MilliporeSigma
D-(−)-Citramalic acid lithium salt ≥95.0% (GC)

D-(−)-Citramalic acid lithium salt

D-(−)-Citramalic acid lithium salt ≥95.0% (GC)
Synonym(s):
(R)-2-Hydroxy-2-methylsuccinic acid lithium salt, D-(−)-2-Methylmalic acid lithium salt
Empirical Formula (Hill Notation):
C5H8O5 · xLi+
CAS Number:
Molecular Weight:
148.11 (free acid basis)

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing