Skip to Content
MilliporeSigma
(+)-cis-Diltiazem hydrochloride ≥99% (HPLC)

(+)-cis-Diltiazem hydrochloride

(+)-cis-Diltiazem hydrochloride ≥99% (HPLC)
Synonym(s):
(2S,3S)-(+)-cis-3-Acetoxy-5-(2-dimethylaminoethyl)-2,3-dihydro-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one hydrochloride, CRD-401
Empirical Formula (Hill Notation):
C22H26N2O4S · HCl
CAS Number:
Molecular Weight:
450.98
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing