Skip to Content
MilliporeSigma
(+)-Carvone certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

D-Carvone

(+)-Carvone certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
(S)-(+)-Carvone, (+)-Carvone, (S)-5-Isopropenyl-2-methyl-2-cyclohexenone, p-Mentha-6,8-dien-2-one, (+)-p-Mentha-6,8-diene 2-one
Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Chemical Composition
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Organoleptic Class
Reference Material Type
Special Grade
Technique