Skip to Content
MilliporeSigma
D-α-Tocopherol polyethylene glycol 1000 succinate BioXtra, water soluble vitamin E conjugate

D-α-Tocopherol polyethylene glycol 1000 succinate

D-α-Tocopherol polyethylene glycol 1000 succinate BioXtra, water soluble vitamin E conjugate
Synonym(s):
D-α-Tocopherol polyethylene glycol succinate, TPGS, Vitamin E polyethylene glycol succinate, Vitamin E-TPGS
CAS Number:

Products

Product Category
Brand
Biological Source
Color
Melting Point (°C)
Physical Form
Product Line
Chemical Composition
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing