Skip to Content
MilliporeSigma
D-Aspartic acid ReagentPlus®, 99%

D-Aspartic acid

D-Aspartic acid ReagentPlus®, 99%
Synonym(s):
(R)-(−)-Aminosuccinic acid, (R)-2-Aminosuccinic acid
Linear Formula:
HO2CCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
133.10
EC Number:

Products

Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing