Skip to Content
MilliporeSigma
Cytidine-5′-monophospho-N-acetylneuraminic acid sodium salt ≥85% (HPLC)

Cytidine-5′-monophospho-N-acetylneuraminic acid sodium salt

Cytidine-5′-monophospho-N-acetylneuraminic acid sodium salt ≥85% (HPLC)
Synonym(s):
CMP-NAN, CMP-NANA, CMP-Neu5Ac, CMP-Sialic acid
Linear Formula:
C20H30N4NaO16P
CAS Number:
Molecular Weight:
636.43

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing