Skip to Content
MilliporeSigma
Cyproheptadine hydrochloride sesquihydrate ≥98% (TLC), solid

Cyproheptadine hydrochloride sesquihydrate

Cyproheptadine hydrochloride sesquihydrate ≥98% (TLC), solid
Synonym(s):
4-(5H-Dibenzo[a,d]cyclohepten-5-ylidene)-1-methyl-piperidine hydrochloride hydrate, Piperidine, 4-(5H-dibenzo[a,d]cyclohepten-5-ylidene)-1-methyl-, hydrochloride, hydrate (2:3)
Empirical Formula (Hill Notation):
C21H21N·HCl·1.5H2O
CAS Number:
Molecular Weight:
350.88
EC Number:

Products

Product Category
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Research Area
Markush Class
Markush Group
Showing 1-2 of 2