Skip to Content
MilliporeSigma
Cymoxanil PESTANAL®, analytical standard

Cymoxanil

Cymoxanil PESTANAL®, analytical standard
Synonym(s):
1-(2-Cyano-2-methoxyiminoacetyl)-3-ethylurea
Empirical Formula (Hill Notation):
C7H10N4O3
CAS Number:
Molecular Weight:
198.18
EC Number:

Products

Product Category
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing